Chloropentaammineruthenium(III) chloride (≥33,5% Ru)

Supplier: Thermo Fisher Scientific
Warning

10520.03 10520.06
10520.03EA 240 EUR
10520.03 10520.06
Chloropentaammineruthenium(III) chloride (≥33,5% Ru)
Chloropentaammineruthenium(III) chloride

CAS No.: 18532-87-1 Molecular Formula: Cl3H15N5Ru Formula Weight: 292.58 Physical Form: Micro Crystals Sensitivity: Hygroscopic Appearance: Yellow/orange MDL No.: MFCD00011529

Formula: Cl₃H₁₅N₅Ru
Storage Temperature: Ambient
MDL Number: MFCD00011529
CAS Number: 18532-87-1
EINECS: 242-408-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR