Potassium pentachloronitrosylruthenate(II) (25,8% Ru)

Supplier: Thermo Scientific
Warning

Synonyms: Potassium pentachloronitrosyl ruthenate(II)

10534.03
10534.03EA 246 EUR
10534.03
Potassium pentachloronitrosylruthenate(II) (25,8% Ru)
Potassium pentachloronitrosylruthenate(II)

CAS No.: 14854-54-7 Molecular Formula: K2RuCl5(NO) Formula Weight: 386.55 Physical Form: Crystalline Melting Point: 220° dec. MDL No.: MFCD00049789

Formula: K₂RuCl₅(NO)
MW: 386,55 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00049789
CAS Number: 14854-54-7
EINECS: 238-919-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR