Tetrakis(triphenylphosphine)platinum(0) (≥15,2% Pt)

Supplier: Thermo Fisher Scientific

Synonyms: Tetrakis(triphenylphosphine)platinum

10549.03 10549.06
10549.03EA 132 EUR
10549.03 10549.06
Tetrakis(triphenylphosphine)platinum(0) (≥15,2% Pt)
Tetrakis(triphenylphosphine)platinum (0)

CAS No.: 14221-02-4 Molecular Formula: Pt[P(C6H5)3]4 Formula Weight: 1244.21 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: Yellow Odour: Odourless Solubility: Soluble in benzene MDL No.: MFCD00010014


Hydrosilation and oxidation

Formula: C₇₂H₆₀P₄Pt
MW: 1244.24 g/mol
MDL Number: MFCD00010014
CAS Number: 14221-02-4
EINECS: 238-087-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR