Amersham™ Protran® Supported Nitrocellulose Western Blotting Membranes, Cytiva
Supplier: Cytiva
10600037 10600020 10600051 10600053 10600015 10600016 10600017 10600018 10600019 10600119 10600120
10600037.EA 452 EUR
10600037. 10600020. 10600051. 10600053. 10600015. 10600016. 10600017. 10600018. 10600019. 10600119. 10600120.
Amersham™ Protran® Supported Nitrocellulose Western Blotting Membranes, Cytiva
Transfer Membranes
Amersham™ Protran® Supported is a robust protein blotting membrane, with 0,2 µm pore size, made of reinforced nitrocellulose for multiple reprobings.
  • Optimal for applications when stripping and reprobing is required
  • Versatile, robust, and excellent signal-to-noise ratios
  • High binding capacity
  • Protran® Supported 0,2 NC membrane recommended for detection of small proteins and peptides (
  • Protran® Supported 0,45 µm membrane suitable for proteins of a wide range of molecular weights
Amersham™ Protran® Supported offers excellent sensitivity, resolution, andMore Product Information