Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
Supplier: Cytiva
10600022 10600102 10600060 10600123
10600022.EA 671 EUR
10600022. 10600102. 10600060. 10600123.
Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
Transfer Membranes
Amersham™ Hybond™ LFP 0.2 PVDF is a low fluorescent, hydrophobic membrane, with 0,2 µm pore size, for protein transfer. Amersham™ Hybond™ LFP is optimised for fluorescent detection in Western blotting applications and for use with the Amersham™ ECL Plex™ detection system.
  • Very low background fluorescence resulting in high sensitivity
  • Provides low auto-fluorescence across a wide range of excitation/emission wavelengths
  • Available in popular pre-cut sizes
  • Protein binding capacity >200 μg/cm²
  • Compatible with a variety of blocking agents, transfer buffers,
More Product Information