Ammonium chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Warning

10632.18 10632.30 10632.A1
10632.18EA 103 EUR
10632.18 10632.30 10632.A1
Ammonium chloride ≥99.999% (metals basis), Puratronic®
Ammonium chloride

Purity: 99.999% CAS No.: 12125-02-9 Molecular Formula: NH4Cl Formula Weight: 53.49 Physical Form: Powder Density: ≈1.53 Refractive Index: 1.642 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 340° subl. Merck Reference: 14,509 MDL No.: MFCD00011420

Formula: NH₄Cl
Melting Pt: 340 °C (subl.)
Density: 1.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 14,509
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR