Nickel ≥99.996% (metals basis), powder, Puratronic® -120 mesh (max. 0.05% C)

Supplier: Thermo Fisher Scientific
Danger

10674.14 10674.22 10674.36
10674.14EA 108 EUR
10674.14 10674.22 10674.36
Nickel ≥99.996% (metals basis), powder, Puratronic® -120 mesh (max. 0.05% C)
Nickel

Purity: 99.996% CAS No.: 7440-02-0 Odour: Odourless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 2371 °C (1013 hPa)
Melting Pt: 1455 °C
Density: 8,9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR