Copper(II) chloride dihydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

10698.06 10698.14 10698.22
10698.06EA 43.1 EUR
10698.06 10698.14 10698.22
Copper(II) chloride dihydrate ≥99.999% (metals basis), Puratronic®
Copper (II) chloride dihydrate

Purity: 99.999% CAS No.: 10125-13-0 Molecular Formula: CuCl2.xH2O (x≈2) Formula Weight: 134.45(anhy) Physical Form: Crystalline Aggregates Density: 2.510 Appearance: Blue-green Odour: Odourless pH: 3.0-3.8 Melting Point: 100° dec. Merck Reference: 14,2633 UN No.: 2802 MDL No.: MFCD00149674

Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.510
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 14,2633

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR