Indium(III) nitrate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Indium trinitrate hydrate

10708.06 10708.14 10708.22
10708.06EA 60.6 EUR
10708.06 10708.14 10708.22
Indium(III) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Indium (III) nitrate hydrate

Purity: 99.999% CAS No.: 207398-97-8 Molecular Formula: In(NO3)3.xH2O Formula Weight: 300.83(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Off-white Melting Point: 100° -H2O UN No.: 1477 MDL No.: MFCD00149742

Formula: InN₃O₉·H₂O
Melting Pt: 100 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR