Indium(III) sulphate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific

10711.09 10711.18
10711.09EA 123 EUR
10711.09 10711.18
Indium(III) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Indium(III) sulphate hydrate

Purity: 99.999% CAS No.: 13464-82-9 Molecular Formula: In2(SO4)3.xH2O (x≈6) Formula Weight: 517.83(anhy) Physical Form: Lump/Powder Melting Point: 250° dec. MDL No.: MFCD00149747

Formula: O₁₂In₂S₃·xH₂O
Melting Pt: 250 °C (dec.)
Density: 3,44 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 13464-82-9
EINECS: 236-689-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR