Lead(II) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lead dichloride

10722.14 10722.22
10722.14EA 159 EUR
10722.14 10722.22
Lead(II) chloride ≥99.999% (metals basis), Puratronic®
Lead (II) chloride

Purity: 99.999% CAS No.: 7758-95-4 Molecular Formula: Cl2Pb Formula Weight: 278.10 Physical Form: Crystalline Density: 5.85 Appearance: White Odour: Odourless Melting Point: 501° Boiling Point: 951° Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157

Formula: Cl₂Pb
MW: 278.11 g/mol
Boiling Pt: 951 °C
Melting Pt: 501 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1 III
Merck Index: 14,5404

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR