Tin ≥99.95% (metals basis), wire, Ø 3.2 mm (0.13 in)

Supplier: Thermo Fisher Scientific

10790.G1 10790.G5
10790.G1EA 119 EUR
10790.G1 10790.G5
Tin ≥99.95% (metals basis), wire, Ø 3.2 mm (0.13 in)
Tin

Purity: 99.95% CAS No.: 7440-31-5 Physical Form: ≈58g/m Odour: Odourless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7,31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR