Ammonium hexachloroosmate(IV) ≥99.99% (metals basis) Os ≥ 42.8%, Premion®

Supplier: Thermo Fisher Scientific
Warning

10823.02 10823.03 10823.06
10823.02EA 0 EUR
10823.02 10823.03 10823.06
Ammonium hexachloroosmate(IV) ≥99.99% (metals basis) Os ≥ 42.8%, Premion®
Ammonium hexachloroosmate(IV)

Purity: 99.99% CAS No.: 12125-08-5 Molecular Formula: H8Cl6N2Os Formula Weight: 438.99 Physical Form: Powder Density: 2.9 Appearance: Brown to black Melting Point: 170° subl. Merck Reference: 14,536 MDL No.: MFCD00010883

Formula: H₈Cl₆N₂Os
Melting Pt: 170 °C (subl.)
Density: 2,9
Storage Temperature: Ambient
MDL Number: MFCD00010883
CAS Number: 12125-08-5
EINECS: 235-188-5
Merck Index: 14,536

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR