Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®

Supplier: Thermo Fisher Scientific
Danger

10824.03 10824.06 10824.14
10824.03EA 143 EUR
10824.03 10824.06 10824.14
Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®
Diammonium tetrachloropalladate

Purity: 99.998% CAS No.: 13820-40-1 Molecular Formula: H8Cl4N2Pd Formula Weight: 284.31 Physical Form: Powder Density: 2.17 Appearance: Dark green to brown Melting Point: dec. MDL No.: MFCD00010884

Formula: H₈Cl₄N₂Pd
Melting Pt: Dec.
Density: 2,17
Storage Temperature: Ambient
MDL Number: MFCD00010884
CAS Number: 13820-40-1
EINECS: 237-498-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR