Perchloric acid ≥70% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

10983.AA 10983.AD 10983.AK
10983.AAEA 52.3 EUR
10983.AA 10983.AD 10983.AK
Perchloric acid ≥70% (metals basis)
Perchloric acid

Purity: 99.9985% CAS No.: 7601-90-3 Molecular Formula: HClO4 Formula Weight: 100.46 Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.670 Appearance: Colourless Odour: Odourless Melting Point: -18° Boiling Point: 203° Merck Reference: 14,7153 UN No.: 1873 MDL No.: MFCD00011325

Formula: HClO₄
Boiling Pt: 203 °C
Melting Pt: –18 °C
Density: 1,670
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
Merck Index: 14,7153
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR