Sodium acetate, anhydrous ≥99.997% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

11005.06 11005.14 11005.22
11005.06EA 49.2 EUR
11005.06 11005.14 11005.22
Sodium acetate, anhydrous ≥99.997% (metals basis)
Sodium acetate

Purity: 99.997% CAS No.: 127-09-3 Molecular Formula: NaOOCCH3 Formula Weight: 82.03 Physical Form: Powder Density: 1.528 pH: 7,5-9 Melting Point: 324° Beilstein Reference: 3595639 Merck Reference: 14,8571 MDL No.: MFCD00012459

Formula: H₃CCOONa
Boiling Pt: 881,4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1,528
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 14,8571

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR