Safety spectacles, IRI-s
Supplier: Bollé Safety
IRIPSI IRIDPSI2.5 IRIPSF IRIDPSI1.5 IRIDPSI3 IRIDPSI2
111-1751EA 13.64 EUR
111-1751 111-1756 111-1750 111-1753 111-1755 111-1754
Safety spectacles, IRI-s
Safety Glasses
These universal safety glasses are ultra-modular and adaptable to every individual thanks to the B-Flex shape memory bridge.
  • Pivoting, non-slip and ergonomic temples
  • Soft B-Flex bridge is fully adjustable and perfectly fits all faces thanks to its shape-memory material
  • Anti-fog coating (on both sides of the lenses) prevents fogging in the most challenging conditions
  • Also available with dioptric corrective lenses
  • Available with or without reading area (clear version only)
Weight: 27 g
Certifications: EN 166, EN 170