VWR® London, Safety Spectacles
Supplier: VWR Collection
111-1845EA 10.7 EUR
111-1845
VWR® London, Safety Spectacles
Safety Glasses
Stylish protective eyewear combining fashion, fit and function.
  • Moulded-in nose bridge
  • Soft, non slip rubber temple tips
  • Ideal for small facial contours
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)