Visors, uvex pheos visor
Supplier: UVEX
9906.002 9906.003 9906.005 9906.006 9906.007 9906.008
111-1935EA 102 EUR
111-1935 111-1936 111-1937 111-1938 111-1939 111-1940
Visors, uvex pheos visor
Face Shields
Polycarbonate visor with uvex supravision excellence coating technology: Permanently anti-fog on the inside and extremely scratch resistant on the outside. Ideal for workplaces where you frequently move between indoor and outdoor environments, from light to dark.
  • Available with two alternative mountings: Mechanical side piece or automatic magnetic lock
  • Compatible with the uvex pheos E-S-WR and uvex pheos B-S-WR safety helmets, as well as with the uvex pheos alpine safety helmet for working at heights
  • Can be combined with uvex K2H and uvex K2H magnet
More Product Information