Safety spectacles, Solus™ 1000
Supplier: 3M
S11SGAF S1SGAFKT S12SGAF S22SGAF S21SGAF S13SGAF S23SGAF S1201SGAF-TSKT S1101NSGAFTSKT
111-2373EA 29 EUR
111-2373 111-2376 111-2374 111-2377 111-2626 111-2375 111-2378 MMMAS1201SGAF-TSKT MMMAS1101NSGAFTSKT
Safety spectacles, Solus™ 1000
Safety Glasses
3M™ Solus™ 1000 series safety glasses feature a strong polycarbonate lens in a slim, stylish frame. These glasses feature a Scotchgard™ Anti-fog/anti-scratch coating, and a soft temple and nose bridge.
  • Stylish and modern slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • Scotchgard™ Anti-Fog coating offers superior anti-fog and anti-scratch properties, even after washing multiple times
  • Lightweight with soft cushioning on nose bridge
  • Suitable for automatic cleaning, disinfection, and sterilisation by
More Product Information