Barium acetate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Warning

11126.06 11126.14 11126.22
11126.06EA 96.6 EUR
11126.06 11126.14 11126.22
Barium acetate ≥99.999% (metals basis), Puratronic®
Barium acetate

Purity: 99.999% CAS No.: 543-80-6 Molecular Formula: C4H6BaO4 Formula Weight: 255.43 Physical Form: Crystalline Density: 2.47 Appearance: White Odour: Odourless Beilstein Reference: 3693411 Merck Reference: 14,965 UN No.: 1564 MDL No.: MFCD00012447

Formula: (H₃CCOO)₂Ba
Melting Pt: ∼450 °C
Density: 2,47
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR