Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)

Supplier: Thermo Fisher Scientific

11144.06 11144.14 11144.22
11144.06EA 150 EUR
11144.06 11144.14 11144.22
Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999+% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937,2 °C
Density: 5,323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR