Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific

Synonyms: Diytterbium trioxide, Ytterbium oxide

11191.14 11191.22 11191.A1
11191.14EA 110 EUR
11191.14 11191.22 11191.A1
Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium (III) oxide

Purity: 99.9% CAS No.: 1314-37-0 Molecular Formula: Yb2O3 Formula Weight: 394.08 Physical Form: Powder Density: 9.2 Melting Point: 2227° Merck Reference: 14,10105 MDL No.: MFCD00011290

Formula: O₃Yb₂
Melting Pt: 2227 °C
Density: 9,2
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0
Merck Index: 14,10105

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR