Examination gloves, nitrile, Micro-Touch® Nitra-Tex™ and Nitra-Tex™ EP™
Supplier: ANSELL MEDICAL PRODUCTS
700124 700123 700122 700121 700115 700114 700113 700112 700111 700125
112-1853EA 22.4 EUR
112-1853 112-1850 112-1849 112-1848 112-1847 112-1846 112-1845 112-1844 112-1843 112-1854
Examination gloves, nitrile, Micro-Touch® Nitra-Tex™ and Nitra-Tex™ EP™
Gloves
NitraTex® gloves are ideal for use in daily examinations and laboratory work in doctors' surgeries, laboratories and hospitals. At 300 mm, Micro-Touch® Nitra-Tex™ EP™ offers additional length for extra protection.
  • Non sterile, powder-free, micro-textured finish
  • 100% nitrile formulation: free from latex proteins, preventing type I (latex allergy) and type IV allergies
  • Strong resistance level to a large number of chemical products, including cytostatics
  • Better puncture resistance than vinyl and most latex gloves
  • Thermoelastic material fits any hand
More Product Information