Cut resistant gloves, ChainExtra
Supplier: HONEYWELL SAFETY
2542000R0302 2542001R0302 2542002R0302 2542005R0302 2542006R0302
112-2894EA 170 EUR
112-2894 112-2895 112-2896 112-2899 112-2900
Cut resistant gloves, ChainExtra
Gloves
These gloves, made of a stainless steel chainmail with detachable plastic straps, provide protection against cuts, a common hazard in meat, fish and processing industries. The gloves are also suitable for use in metal industries.
  • Detachable and interchangeable plastic straps with antimicrobial agent
  • Firm fastening (press studs) for a secure fit
  • Without side slit which may allow a knife to enter at wrist level
  • Fitted with an identification tag which can be personalised by the wearer
  • Exceptional quality of chain mesh welds, reducing the number of repairs,
More Product Information