Knitted gloves, BO600 / BO605
Supplier: Showa
BO605-S BO605-M BO605-L BO605-XL BO600-S BO600-M BO600-L BO600-XL BO605-XXL
112-4221EA 50.5 EUR
112-4221 112-4220 112-4219 112-4222 112-4311 112-4310 112-4309 112-4312 112-4223
Knitted gloves, BO600 / BO605
Gloves
These seamless knitted nylon gloves with elasticated cuffs feature a PU coating on the fingertips. The coating protects the hand from oils and micro cuts.
  • Lightweight, elastic, low linting
  • Anatomical shape, resistant to deformation
  • No fingerprints left on handled objects
  • Minimal allergy risk, no irritation by seams
  • Available in different lengths: Long version provides protection for the wrist
Ideal for light assembly tasks in the automotive and metalworking industry as well as in logistics.
Colour: White
EN 388 (0131); Cat. II
Certifications:More Product Information