Nitrile gloves, Microflex® 93-833
Supplier: Ansell
93-833/8.5-9 93-833/6.5-7 93-833/7.5-8 93-833/5.5-6 93-833/9.5-10
112-4273EA 41.7 EUR
112-4273 112-4271 112-4272 112-4270 112-4274
Nitrile gloves, Microflex® 93-833
Gloves
These thin disposable gloves are up to 60% stronger than leading nitrile brands and up to 90% stronger than leading latex brands. They feature an advanced proprietary nitrile formulation which delivers an increased tactile sensitivity and reliable durability.
  • Designed to reduce hand fatigue and help workers be more productive while exerting less muscle effort
  • Help to protect workers from Carpal Tunnel Syndrome and minimise the risk factors towards developing ergonomic work place injuries
  • Contain no natural rubber latex or powder to help protect wearers
More Product Information