Industrial gloves, oil repellent, HyFlex® 11-920
Supplier: Ansell
11-920/8 11-920/10 11-920/11 11-920/6 11-920/7 11-920/9
112-4288EA 125 EUR
112-4288 112-4284 112-4285 112-4286 112-4287 112-4289
Industrial gloves, oil repellent, HyFlex® 11-920
Gloves
This nylon liner with nitrile palm coating features Ansell Grip Technology™. Microscopic channels in the glove coating guide away the oil-resulting in a secure glove-to-surface contact. This allows the wearer to securely handle small and medium sized oily components.
  • Protects skin from oil contamination
  • Excellent grip in oily environments
  • Outstanding fit and comfort, excellent dexterity and tactility
  • Anti-static according to EN 1149
Ideal for assembly and finishing tasks, quality control, metal fabrication and handling as well as factory maintenance.
More Product Information