General purpose gloves, TEGERA® 888
Supplier: EJENDALS
888-10 888-11 888-12 888-6 888-7 888-8 888-9
112-5380EA 0 EUR
112-5380 112-5381 112-5382 112-5383 112-5384 112-5385 112-5386
General purpose gloves, TEGERA® 888
Gloves
These unlined gloves are made of a combination of full grain cowhide leather (50%), cotton (49%) and natural latex (1%).
  • Reinforced index finger, reinforced fingers and thumb
  • Elasticated 360° fastening
  • High level of protection with good fingertip sensitivity
  • Durable and good fit
  • Ideal for works with medium weight
The gloves prevent risks of abrasion injuries, blisters, grazes, scratches, lacerations and contact with dirt.
Primary areas of use: Airport work, assembly, cleaning, building and construction, warehouse work, carpentry, installation work,More Product Information