General purpose gloves, Showa 370
Supplier: Showa
370-M 370-S 370-XL 370-XS 370-XXL 370-L
112-5798EA 81.2 EUR
112-5798 112-5799 112-5800 112-5801 112-5802 112-5797
General purpose gloves, Showa 370
Gloves
These lightweight palm-coated nylon gloves are ideal for protecting hands against fluids such as oils. They allow finger dexterity to be maintained whilst being robust and hard-wearing.
  • Nitrile coating protects the hand from oil, hydrocarbon and grease penetration
  • Breathable back of hand and fingers
  • High levels of dexterity and tactility
  • Seamless 13 gauge lint-free liner provides excellent dry grip and resistant to deformation
Applications include assembling components, handling greasy components, inspection, machining and maintenance.These gloves areMore Product Information