Impact resistant gloves, showa 377-IP
Supplier: Showa
377IP-10/XXL 377IP-7/M 377IP-8/L 377IP-9/XL
112-5813EA 198 EUR
112-5813 112-5814 112-5815 112-5816
Impact resistant gloves, showa 377-IP
Gloves
These nitrile foam Hi-Vis anti-impact gloves have a highly protective design to prevent injuries caused by impact and other hazards. The SHOWA-configured Thermo Plastic Rubber (TPR) shields to the back of the fingers and hand are designed to bolster protection against crushes and impacts.
  • Designed for easy movement and continuous wear
  • No irritation by seams in contact with the skin
  • Well protected wrist
  • Anatomical shape
  • Reinforced knit cuff
The SHOWA patented nitrile foam coating provides excellent grip in wet and greasy conditions, flat nitrileMore Product Information