Chemical and cold resistant gloves, Showa 465
Supplier: Showa
465-8/M 465-9/L 465-10/XL
112-5847EA 29 EUR
112-5847 112-5848 112-5846
Chemical and cold resistant gloves, Showa 465
Gloves
These flexible and soft gloves feature a removable, insulated acrylic liner and a full PVC coating for oil and chemical resistance.
  • Absorb perspiration: For comfortable extended wear
  • Seal and protect the hand against chemicals while remaining flexible, up to −20 °C
  • Impermeable for working in damp or greasy environments
  • Rough finish over entire surface for better grip
  • Effective cold protection, with easy clean lining
  • Extended protection on the forearm
Typical applications include: Public utilities, logistics, agriculture, transport, petrochemical,More Product Information