Life science nitrile gloves, PUREZERO® MARIN®
Supplier: Halyard
LFS111LG LFS111MD LFS111SM LFS111XS LFS111XL
112-6268EA 23.4 EUR
112-6268 112-6267 112-6266 112-6265 112-6269
Life science nitrile gloves, PUREZERO® MARIN®
Gloves
PUREZERO® life science nitrile gloves deliver on all fronts. They are tested against 57 chemicals, with 3 levels of colour-coded protection offering increasing thickness designed for life science applications. The SAFESKIN® manufacturing facility uses 92% renewable energy, dispensers are made with up to 85% recycled materials, and cases are made with up to 99% recycled pulp.
  • Barrier protection (57 chemicals, 14 chemotherapy drugs)
  • Good fit and comfort
  • Textured fingertips provide tactile agility
  • Beaded cuff allows for ease of donning
  • Not made with natural
More Product Information