Samarium trichloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11232.06 11232.14 11232.22
11232.06EA 54.9 EUR
11232.06 11232.14 11232.22
Samarium trichloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium trichloride

Purity: 99.99% CAS No.: 10361-82-7 Molecular Formula: SmCl3.xH2O Formula Weight: 256.71(anhy) Physical Form: Wet Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8347 MDL No.: MFCD00011235

Formula: Cl₃Sm·xH₂O
Melting Pt: 110 °C (-5H₂O)
Density: 2,383 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011235
CAS Number: 10361-82-7
EINECS: 233-797-0
Merck Index: 14,8347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR