Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11264.30 11264.18 11264.A1
11264.30EA 197 EUR
11264.30 11264.18 11264.A1
Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) oxide

Purity: 99.99% CAS No.: 1312-81-8 Molecular Formula: La2O3 Formula Weight: 325.82 Physical Form: Powder Density: 6.51 Melting Point: 2307° Boiling Point: 4200° Merck Reference: 14,5363 MDL No.: MFCD00011071

Formula: La₂O₃
Boiling Pt: 4200 °C
Melting Pt: 2307 °C
Density: 6,51
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR