Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Europium (III) nitrate hydrate

11296.06 11296.14 11296.02
11296.06EA 274 EUR
11296.06 11296.14 11296.02
Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium trinitrate hydrate

Purity: 99.99% CAS No.: 100587-95-9 Molecular Formula: Eu(NO3)3.xH2O (x≈6) Formula Weight: 337.97(anhy) Physical Form: Crystalline Aggregates Appearance: Colourless to pink Melting Point: 85° Merck Reference: 14,3904 UN No.: 1477 MDL No.: MFCD00149700

Formula: EuN₃O₉·xH₂O
Melting Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00149698
CAS Number: 100587-95-9
EINECS: 233-380-3
UN: 1477
ADR: 5.1,III
Merck Index: 14,3904

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR