VWR® Basic Cleanroom Overalls, SMS
Supplier: VWR Collection
113-1097EA 194 EUR
113-1097 113-1103 113-1102 113-1101 113-1099 113-1096 113-1098 113-1095 113-1094 113-1093 113-1092 113-1091 113-1090
VWR® Basic Cleanroom Overalls, SMS
Disposable Garments
These cleanroom overalls are manufactured from a latex-free, multi-layer spun-bonded - meltblown - spun-bonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
  • Tapered collar, inset sleeves, zip closure, elasticated cuffs
  • Fluid and particulate barrier
  • Light chemical splash resistance
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Cleanroom,Laboratory