Chemical resistant overalls, Tyvek® 500 Xpert, models CHF5 / CHF5a
Supplier: DuPont
TYCHF5SGR2X010 TYCHF5SBUSM010 TYCHF5SGRSM010 TYCHF5SGRMD010 TYCHF5SGRLG010 TYCHF5SGRXL010 TYCHF5SBUMD010 TYCHF5SBULG010 TYCHF5SBUXL010 TYCHF5SBU2X010 TYCHF5SWHLG01P TYCHF5SWHSM01P TYCHF5SWHMD01P TYCHF5SWHXL01P TYCHF5SWH2X01P TYCHF5SWH3X01P TYCHF5SWH2X01B TYCHF5SWHXL01B TYCHF5SWHSM01B TYCHF5SWH3X01B TYCHF5SWHMD01B TYCHF5SWHLG01B TYCHF5SBU3X010 TYCHF5SGR3X010 TYCHF5SWH4X01P TYCHF5SWH5X01P TYCHF5SWH6X01P TYCHF5SWH7X01P TYCHF5SGR00
113-3868EA 422 EUR
113-3868 113-3869 113-3864 113-3865 113-3866 113-3867 113-3870 113-3871 113-3872 113-3873 113-6062 113-6060 113-6061 113-6063 113-6064 113-6065 113-8343 113-8342 113-8339 113-8344 113-8340 113-8341 113-9682 113-9683 DUPOTYCHF5SWH4X01P DUPOTYCHF5SWH5X01P DUPOTYCHF5SWH6X01P DUPOTYCHF5SWH7X01P DUPOTYCHF5SGR00
Chemical resistant overalls, Tyvek® 500 Xpert, models CHF5 / CHF5a
Disposable Garments
These Tyvek® overalls are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Ultra-low linting fabric is robust yet lightweight (<180 g per garment)
  • 3-piece hood for optimal fit to head and face when turning
  • Large, 'easy to grasp' zip pull
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
More Product Information