Chemical Resistant Overalls, 4515
Supplier: 3M
4515W3XL 4515WXL 4515WL 4515WM 4515B3XL 4515OM 4515OS 4515OL 4515O3XL 4515O2XL 4515BXL 4515BS 4515BM 4515B4XL 4515B2XL 4515BL 4515OXL 4515WS 4515W4XL 4515WXXL
113-6275EA 197 EUR
113-6275 113-6280 113-6277 113-6278 113-6255 113-6264 113-6265 113-6263 113-6262 113-6261 113-6260 113-6259 113-6258 113-6256 113-6254 113-6257 113-6266 113-6279 113-6276 MMMA4515WXXL
Chemical Resistant Overalls, 4515
Disposable Garments
These disposable overalls made of highly breathable SMS material provide basic protection against dry particles, certain liquid chemical splashes and hazardous dusts such as asbestos fibres.
  • Breathable material technology for reduced heat stress
  • Robust, low linting
  • Two-way zip with storm flap (incl. tape tabs) for convenience and additional protection
  • Elasticated cuffs, waist and ankles for a better fit and freedom of movement
Typical light duty applications may include asbestos inspection, insulation laying, cement manufacturing and metal grindingMore Product Information