High visibility rain jacket, Brighton 132Z
Supplier: SIOEN
132Z/415/S 132Z/415/M 132Z/415/L 132Z/415/1XL 132Z/415/2XL 132Z/415/3XL 132Z/414/S 132Z/414/M 132Z/414/L 132Z/414/1XL 132Z/414/2XL 132Z/414/3XL 132ZA2EJ4415XS
113-7701EA 228 EUR
113-7701 113-7700 113-7699 113-7696 113-7697 113-7698 113-7695 113-7694 113-7693 113-7690 113-7691 113-7692 SIOE132ZA2EJ4415XS
High visibility rain jacket, Brighton 132Z
Workwear
Waterproof and windproof rain jacket made of Siopor® Extra: 100% polyester fabric (cotton touch) with 100 % PU laminate. The fabric is water repellent on the outside and moisture attracting on the inside. With fixed mesh lining in body and hood and polyamide lining in sleeves.
  • Straight collar, detachable hood with chin protection and peak
  • Zip closure under double flap with press studs
  • Two inset chest pockets with zip closure, two patched pockets with zip closure, Napoleon pocket, two inside pockets
  • Round inset sleeves, narrowing by touch and close
More Product Information