Quantus®, Underwear Sweatshirts
Supplier: VWR Collection
113-9176EA 0 EUR
113-9176 113-9169 113-9170 113-9179 113-9173 113-9177
Quantus®, Underwear Sweatshirts
Workwear
The Quantus® underwear sweatshirt is made from a high-quality breathable polyester microfibre material with carbon. The fabric was specially developed for use as underwear in combination with a cleanroom overall or coat. It serves as a prefilter to allow open-pore-fabric to be used in a higher cleanroom class.
  • Sweatshirt with long sleeves and round neck
  • Extra material flap for bar code inside
  • Soft grip and greater comfort when wearing
  • Anti-static and antimicrobial protection
  • Recommended for wear underneath disposable and reusable clothing
  • RFID chip
More Product Information