Erbium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11312.09 11312.18
11312.09EA 191 EUR
11312.09 11312.18
Erbium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trifluoride

Purity: 99.99% CAS No.: 13760-83-3 Molecular Formula: ErF3 Formula Weight: 224.28 Physical Form: Granular/Powder Density: 6.4 Melting Point: 1350° MDL No.: MFCD00016074

Formula: ErF₃
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1350 °C
Density: 6,4
Storage Temperature: Ambient
MDL Number: MFCD00016074
CAS Number: 13760-83-3
EINECS: 237-356-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR