Safety ankle boots, lace-up, The Roar, Predator
Supplier: U-Power
SN10044/35 SN10044/36 SN10044/37 SN10044/38 SN10044/39 SN10044/40 SN10044/41 SN10044/42 SN10044/43 SN10044/44 SN10044/45 SN10044/46 SN10044/47 SN10044/48
114-2878EA 121 EUR
114-2878 114-2879 114-2880 114-2881 114-2882 114-2883 114-2884 114-2885 114-2886 114-2887 114-2888 114-2889 114-2890 114-2891
Safety ankle boots, lace-up, The Roar, Predator
Shoes and Boots
Safety ankle boots made from soft water repellent leather 'antique style' and a slip-resistant dual-density PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Grey
Order Now
SPECIFICATIONS
Environmentally Preferable