ESD safety ankle boots, lace-up, Safety-Grip, Renzo XW Mid
Supplier: ELTEN
765861/36 765861/37 765861/38 765861/39 765861/40 765861/41 765861/42 765861/43 765861/44 765861/45 765861/46 765861/47 765861/48 765861/49 765861/50
114-8162EA 68.9 EUR
114-8162 114-8163 114-8164 114-8165 114-8166 114-8167 114-8168 114-6999 114-8169 114-8170 114-8171 114-8172 114-8173 114-8174 114-8175
ESD safety ankle boots, lace-up, Safety-Grip, Renzo XW Mid
Shoes and Boots
Durable ESD safety shoes made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Full length fleece inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
  • Extra wide version (XW)
Ideal for use in areas whereMore Product Information