Silver(I) sulphide ≥99%

Supplier: Thermo Fisher Scientific
Warning

11416.06 11416.14 11416.22
11416.06EA 61.4 EUR
11416.06 11416.14 11416.22
Silver(I) sulphide ≥99%
Silver (I) sulphide

Purity: 99% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Powder Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Solubility: Soluble in HNO3, alkali cyanide solutions. Insoluble in water Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formula: Ag₂S
Melting Pt: 825 °C
Density: 7,317
Storage Temperature: Ambient
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
ADR: 9,III
Merck Index: 14,8530

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR