Diethyl malonate 99+%

Supplier: Thermo Fisher Scientific

114450010 114450250 114455000 114450025 114450100
114450010.EA 106 EUR
114450010. 114450250. 114455000. 114450025. 114450100.
Diethyl malonate 99+%
Diethyl malonate
Formula: C₇H₁₂O₄
Boiling Pt: 199 °C (760 mmHg)
Melting Pt: –50 °C
Density: 1,055 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20°C, 589 nm)
Density (20°C) 1.052 to 1.058 g/ml
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR