Lab soakers, Versi-Dry® and Super Versi-Dry®, Nalgene®
Supplier: Thermo Fisher Scientific
62050-00 62060-00 62065-00 62070-00 62080-00 74000-00 74018-00 74040-00 74043-00 74050-00 74200-00 74218-00 74240-00
115-0011EA 266 EUR
115-0011 115-9257 115-9258 115-9259 115-9260 115-9267 115-9268 115-9269 115-0012 115-0013 115-0014 115-0015 115-0016
Lab soakers, Versi-Dry® and Super Versi-Dry®, Nalgene®
Surface Protectors Bench Protectors
Versi-Dry lab soakers protect surfaces and equipment, cushion breakable items and quickly absorb spills. The polythene backing is waterproof and resistant to chemicals. Standard version absorbs 750 ml/m², super version absorbs 1050 ml/m².
  • Multiple layers help protect glassware and other delicate items from breaking when dropped or tipped over on hard surfaces
  • Soft and non abrasive, does not scratch delicate surfaces or equipment
  • Can also be used as a tray, drawer or shelf liner or to wipe up spills