Mop systems, Edgeless®
Supplier: CONTEC
EL-MOP EL-MOPIR 2646F
115-0174EA 64 EUR
115-0174 CONTEL-MOPIR CONT2646F
Mop systems, Edgeless®
Mops
Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.
  • Mop heads are free from any cellulose contaminants
  • Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
  • All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation