Hand cleansing cream, Kresto® classic
Supplier: Deb-STOKO
87147A06 KCL250ML KCL2LT
115-1509EA 203 EUR
115-1509 115-2293 115-2294
Hand cleansing cream, Kresto® classic
Skincare Soaps and Skin Cleansers
Hand cleanser paste for extra heavy-duty contamination from, for example, oils, greases, drilling mud, tar, carbon black, bitumen, graphite and metallic dust lubricants.
  • Contains toxicologically safe solvents to lift difficult to remove oil-based contaminants without harming the skin
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • Contains Eucornol® skin protectant to minimise the risk of skin irritation
  • Non soapy, petroleum solvent-free, silicone-free, colourant-free, perfumed