Cleaning wipes
Supplier: MICRONOVA
WP4-99IR WP2-1212 WP2-99 WP6-99H WPAA-1212 WP20-1212 WP20-99 WP4-99 WP68-99H WPAA-99
115-1879EA 3150 EUR
115-1879 MICRWP2-1212 MICRWP2-99 MICRWP6-99H MICRWPAA-1212 MICRWP20-1212 MICRWP20-99 115-1878 MICRWP68-99H 129-3812
Cleaning wipes
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
A range of absorbent and low-linting wipes to meet every cleaning, sanitizing, polishing and drying application in the cleanroom, support areas, and research suites.
  • Made with cleanroom approved materials
  • Durable, autoclavable, reusable and absorbent
  • Choose from a range of fabrics for specialized cleaning
  • Ideal for parts and surface cleaning, removing tough soils, liquid removal
  • Following laundering, wipes can be used in less critical areas
  • Two-ply wipes feature an inverted hem
  • Hold up to strong disinfectants
NovaCel™ Wipes: Ultra-clean PolyvinylMore Product Information